Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01295946_m1
          See other CFHR1 GE Assays ›
          Gene Symbol:
          Cfhr1
          Gene Name:
          complement factor H-related 1
          Gene Aliases:
          AI194696, CFHRB, Cfhl1
          Chromosome Location:
          Chr.1: 139547064 - 139560222 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01295946_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cfhr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_015780.2 NP_056595.1 2 - 3 330 96
          GenBank mRNA BC111870.1 - 2 - 3 313 40057075 96
          M29008.1 - 2 - 3 330 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003186.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cfhr1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513460 BC111870.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513460 M29008.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513460 NM_015780.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513460 BC111870.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513460 M29008.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513460 NM_015780.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648553 AK007095.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930626 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 291991 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 350136 AK077182.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905557 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 482383 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920676 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 6355 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 6151 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118476 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159545 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513460 BC111870.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513460 M29008.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513460 NM_015780.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513460 BC111870.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513460 M29008.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513460 NM_015780.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20663512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20663520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20663521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20663519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20663515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20663527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20663523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20663518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20663511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20663514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20663528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20663524 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118476 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159545 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00200826 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04284571 - -
          Whole Mouse Genome G4122A A_51_P513460 BC111870.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513460 M29008.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513460 NM_015780.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513460 BC111870.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513460 M29008.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513460 NM_015780.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117934 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105124 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159545 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118476 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00054708 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00575774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01127586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00575772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00259507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01127590 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10358322 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358323 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358320 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358316 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358319 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358317 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358318 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358321 - -
          Affymetrix HT MG-430 PM Array Plate 1419436_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10358299 - -
          Affymetrix Mouse Gene 1.1 ST Array 10358315 - -
          Mouse Genome 430 2.0 Array 1419436_PM_at - -
          Mouse Genome 430 2.0 Array 1419436_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 50702_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 50702_at - -
          Affymetrix Mouse Clariom S Array PSR0100025827.mm.1 NM_015780.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025827.mm.1 NM_015780.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025827.mm.1 TC0100003186.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025833.mm.1 NM_015780.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025833.mm.1 NM_015780.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025833.mm.1 TC0100003186.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025830.mm.1 NM_015780.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025830.mm.1 NM_015780.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025830.mm.1 TC0100003186.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025834.mm.1 NM_015780.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025834.mm.1 NM_015780.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025834.mm.1 TC0100003186.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025832.mm.1 NM_015780.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025832.mm.1 NM_015780.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025832.mm.1 TC0100003186.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025828.mm.1 NM_015780.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025828.mm.1 NM_015780.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025828.mm.1 TC0100003186.mm.1 MTA TC
          Mouse Expression Set 430 1419436_PM_at - -
          Mouse Expression Set 430 1419436_at - -
          Murine Genome U74v2 Set 92291_PM_f_at - -
          Murine Genome U74v2 Set 92291_f_at - -
          Mouse Genome 430A 2.0 Array 1419436_at - -
          Mouse Genome 430A 2.0 Array 1419436_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419436_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419436_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 475027 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1850050 - -
          Illumina mouse-6 v1.0 expression beadchip 5550519 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001850050 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005550519 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2735449 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2720352 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2735449 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2720352 - -
          mouse-6 v1.1 (Illumina) scl050702.1_126-S - -
          mouse-6 v1.1 (Illumina) scl16210.3.1_71-S - -
          Illumina Sentrix mouse-6.v1 bead array 1850050 - -
          Illumina Sentrix mouse-6.v1 bead array 5550519 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl16210.3.1_71-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl050702.1_126-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1850050 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5550519 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2735449 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2881864 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2720352 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2735449 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2720352 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2881864 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline