Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01296050_m1
          See other ANKRD45 GE Assays ›
          Gene Symbol:
          Ankrd45
          Gene Name:
          ankyrin repeat domain 45
          Gene Aliases:
          4933409K03Rik
          Chromosome Location:
          Chr.1: 161142547 - 161170507 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01296050_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ankrd45
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028664.1 NP_082940.1 6 - 7 922 69
          GenBank mRNA AK016757.1 - 5 - 6 841 69
          BC049713.1 - 6 - 7 894 6741553 69
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AK144074.1 - 1401 Chr. - - [-] on Build 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001466.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          kinase inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P214772 NM_028664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P214772 AK016757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P214772 BC049713.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P214772 NM_028664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P214772 AK016757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P214772 BC049713.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 141781 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 594626 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 676818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667174 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60813 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 213405 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 958511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 708835 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351048 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26389 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 756343 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 65254 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 664169 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487851 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 950177 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 595329 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127858 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 137255 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 140219 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162705 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159677 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P214772 NM_028664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P214772 AK016757.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P214772 BC049713.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P214772 NM_028664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P214772 AK016757.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P214772 BC049713.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162705 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159677 - -
          Whole Mouse Genome G4122A A_52_P214772 NM_028664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P214772 AK016757.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P214772 BC049713.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P214772 NM_028664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P214772 AK016757.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P214772 BC049713.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P214770 NM_028664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P214770 AK016757.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P214770 BC049713.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162705 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20770277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01023874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20770258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20770290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00178775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20770274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00372134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20770275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20770266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01041465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20770255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20770252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20770297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20770280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01023875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01023871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00372139 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20770294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20770267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00386478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00372122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00995398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00374851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00880761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00372133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20770295 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10351061 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351057 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351059 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351060 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351062 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351058 - -
          Affymetrix HT MG-430 PM Array Plate 1430395_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10351056 - -
          Mouse Genome 430 2.0 Array 1430395_at - -
          Mouse Genome 430 2.0 Array 1430395_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73844_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73844_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100012084.mm.1 NM_028664.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012084.mm.1 NM_028664.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012084.mm.1 TC0100001466.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100012085.mm.1 NM_028664.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012085.mm.1 NM_028664.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012085.mm.1 TC0100001466.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100012083.mm.1 NM_028664.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012083.mm.1 NM_028664.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012083.mm.1 TC0100001466.mm.1 MTA TC
          Mouse Expression Set 430 1430395_at - -
          Mouse Expression Set 430 1430395_PM_at - -
          Murine Genome U74v2 Set 168252_f_at - -
          Murine Genome U74v2 Set 168252_PM_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1430395_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430395_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 403281 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240591 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240591 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0