Search
Search

Pcbp2| Gene Symbol: Pcbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103165.1 | NP_001096635.1 | 5 - 6 | 485 | 90 | |
| NM_001103166.1 | NP_001096636.1 | 5 - 6 | 485 | 90 | ||
| NM_001174073.1 | NP_001167544.1 | 5 - 6 | 461 | 90 | ||
| NM_011042.2 | NP_035172.2 | 5 - 6 | 485 | 90 | ||
| GenBank mRNA | AK012297.1 | - | 5 - 6 | 333 | 90 | |
| AK161527.1 | - | 5 - 6 | 309 | 90 | ||
| AK169268.1 | - | 5 - 6 | 579 | 90 | ||
| BC020478.1 | - | 4 - 5 | 230 | 90 | ||
| BQ900329.1 | - | 5 - 6 | 461 | 90 | ||
| EU604548.1 | - | 4 - 5 | 240 | 90 | ||
| L19661.1 | - | 5 - 6 | 372 | 90 | ||
Set Membership: |
|