Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01296752_m1
          See other ZFP369 GE Assays ›
          Gene Symbol:
          Zfp369
          Gene Name:
          zinc finger protein 369
          Gene Aliases:
          B930030B22Rik, D230020H11Rik, NRIF2
          Chromosome Location:
          Chr.13: 65276668 - 65297804 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01296752_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp369
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178364.5 NP_848141.3 3 - 4 343 84
          GenBank mRNA AJ319726.1 - 3 - 4 360 84
          AK082535.1 - 3 - 4 346 84
          BC036565.1 - 383 84

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp369

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          apoptotic process
          regulation of cell cycle
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          nucleic acid binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          neurotrophin p75 receptor binding
          protein binding
          sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102609 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102611 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102454 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102610 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102455 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038081 NM_178364.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038081 AJ319726.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P61703 AK084309.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P61703 NM_178364.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P61703 AK084309.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P61703 NM_178364.5 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744494 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 212420 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 161792 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 623305 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 944285 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 663906 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144517 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 208480 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 592965 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11091 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 6601 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137011 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155980 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2038081 NM_178364.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2038081 AJ319726.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P61703 AK084309.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P61703 NM_178364.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P61703 AK084309.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P61703 NM_178364.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P156316 AJ319726.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102616 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07902912 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137011 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155980 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07902915 - -
          Whole Mouse Genome G4122A A_51_P156316 AJ319726.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P61703 AK084309.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P61703 NM_178364.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P61703 AK084309.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P61703 NM_178364.5 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123521 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137011 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155980 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10593145 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10998707 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10998708 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10593138 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10593140 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102455 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102454 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102610 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102609 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10723786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10858087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10521282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10879554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10684322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10523045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10548234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10879547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10548238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10998708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10523046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10467985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10684324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10858080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10838604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10723785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102631 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10405856 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405861 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405858 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405860 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405863 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405859 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405857 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405862 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405855 - -
          Affymetrix HT MG-430 PM Array Plate 1427538_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1455681_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10405853 - -
          Mouse Genome 430 2.0 Array 1455681_at - -
          Mouse Genome 430 2.0 Array 1455681_PM_at - -
          Mouse Genome 430 2.0 Array 1427538_PM_at - -
          Mouse Genome 430 2.0 Array 1427538_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170936_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170936_at - -
          Mouse Expression Set 430 1427538_at - -
          Mouse Expression Set 430 1427538_PM_at - -
          Mouse Expression Set 430 1455681_PM_at - -
          Mouse Expression Set 430 1455681_at - -
          Murine Genome U74v2 Set 137614_PM_at - -
          Murine Genome U74v2 Set 137614_at - -
          Mouse Genome 430A 2.0 Array 1427538_at - -
          Mouse Genome 430A 2.0 Array 1427538_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427538_at - -
          Affymetrix HT Mouse Genome 430A Array 1427538_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455681_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455681_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4070458 - -
          Illumina mouse-6 v1.0 expression beadchip 4070458 - -
          Illumina mouse-6 v1.0 expression beadchip 6860040 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004070458 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006860040 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2508173 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2520663 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2520663 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2508173 - -
          mouse-6 v1.1 (Illumina) scl00170936.1_203-S - -
          mouse-6 v1.1 (Illumina) scl0170936.4_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 6860040 - -
          Illumina Sentrix mouse-6.v1 bead array 4070458 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00170936.1_203-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0170936.4_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6860040 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4070458 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2520663 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2857692 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2857692 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2520663 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00170936.1_203-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline