Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01297406_m1
          See other NOXRED1 GE Assays ›
          Gene Symbol:
          Noxred1
          Gene Name:
          NADP+ dependent oxidoreductase domain containing 1
          Gene Aliases:
          4933437F05Rik
          Chromosome Location:
          Chr.12: 87221123 - 87238774 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01297406_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Noxred1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027744.1 NP_082020.1 6 - 7 869 93
          GenBank mRNA AK017096.1 - 6 - 7 1000 93
          BC125331.1 - 6 - 7 869 40129846 93
          BC125333.1 - 6 - 7 869 40129848 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002119.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          reductase

          Gene Ontology Categories:

          Function(s) Process(es)

          proline biosynthetic process
          oxidation-reduction process
          molecular_function
          pyrroline-5-carboxylate reductase activity
          N-ethylmaleimide reductase activity
          oxidoreductase activity
          reduced coenzyme F420 dehydrogenase activity
          sulfur oxygenase reductase activity
          malolactic enzyme activity
          NADPH:sulfur oxidoreductase activity
          epoxyqueuosine reductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P505662 BC125331.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P505662 BC125333.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P505662 NM_027744.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P505662 AK017096.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P505662 BC125331.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P505662 BC125333.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P505662 NM_027744.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P505662 AK017096.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 757284 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 922205 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 805682 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 710731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297590 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 395143 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897707 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 70991 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 653302 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 573937 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 116520 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 43859 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170301 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119654 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P505662 BC125331.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P505662 BC125333.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P505662 NM_027744.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P505662 AK017096.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P505662 BC125331.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P505662 BC125333.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P505662 NM_027744.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P505662 AK017096.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119654 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02571116 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170301 - -
          Whole Mouse Genome G4122A A_51_P505662 BC125331.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505662 BC125333.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505662 NM_027744.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505662 AK017096.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505662 BC125331.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505662 BC125333.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505662 NM_027744.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505662 AK017096.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170301 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10377811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09933505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10377804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10243085 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10401748 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401745 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401752 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401751 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401749 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401746 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401750 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401744 - -
          Affymetrix HT MG-430 PM Array Plate 1430314_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10401743 - -
          Mouse Genome 430 2.0 Array 1430314_PM_at - -
          Mouse Genome 430 2.0 Array 1430314_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71275_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71275_at - -
          Affymetrix Mouse Clariom S Array PSR1200014693.mm.1 NM_027744.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014693.mm.1 NM_027744.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014693.mm.1 TC1200002119.mm.1 MTA TC
          Mouse Expression Set 430 1430314_at - -
          Mouse Expression Set 430 1430314_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430314_at - -
          Affymetrix HT Mouse Genome 430B Array 1430314_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 475429 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2120113 - -
          MouseWG-6 v2 Expression BeadChip 2630541 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2720236 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226545 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2720236 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226545 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1226545 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline