Search
Search

Morc4| Gene Symbol: Morc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193309.1 | NP_001180238.1 | 2 - 3 | 454 | 76 | |
| NM_029413.4 | NP_083689.2 | 2 - 3 | 454 | 76 | ||
| GenBank mRNA | AK017472.1 | - | 2 - 3 | 423 | 76 | |
| AK032807.1 | - | 2 - 3 | 455 | 76 | ||
| AK048519.1 | - | 2 - 3 | 423 | 76 | ||
| AK053895.1 | - | 2 - 3 | 426 | 76 | ||
| AK164017.1 | - | 2 - 3 | 422 | 76 | ||
| BC098483.1 | - | 2 - 3 | 409 | 30022418 | 76 | |
| BC132153.1 | - | 2 - 3 | 308 | 40130430 | 76 | |
| BC132497.1 | - | 2 - 3 | 308 | 40130774 | 76 | |
Set Membership: |
|