Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01299104_g1
          See other ODF3B GE Assays ›
          Gene Symbol:
          Odf3b
          Gene Name:
          outer dense fiber of sperm tails 3B
          Gene Aliases:
          2010001J22Rik
          Chromosome Location:
          Chr.15: 89377447 - 89379681 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01299104_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Odf3b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013022.1 NP_001013040.1 4 - 5 616 86
          GenBank mRNA AK008010.1 - 3 - 4 508 86
          BC087962.1 - 4 - 5 616 6748887 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500002053.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Odf3b

          Panther Classification:

          Molecular Function -

          structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424874 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424876 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424872 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424871 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424873 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422369 NM_001013022.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422369 AK008010.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422369 BC087962.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422369 NM_001013022.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422369 AK008010.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422369 BC087962.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422369 NM_001013022.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422369 AK008010.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422369 BC087962.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422369 NM_001013022.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422369 AK008010.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422369 BC087962.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424904 - -
          Whole Mouse Genome G4122A A_51_P422369 NM_001013022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422369 AK008010.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422369 BC087962.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422369 NM_001013022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422369 AK008010.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422369 BC087962.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11943656 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12061271 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11943640 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12061275 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11943653 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11943654 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424876 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424874 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424871 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424872 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12260943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11943653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12002372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12403995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11943650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12083516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11943646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12083522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12083521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424922 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10431563 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431561 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431562 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431559 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431560 - -
          Affymetrix HT MG-430 PM Array Plate 1439666_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1443671_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1443670_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10431558 - -
          Mouse Genome 430 2.0 Array 1443671_x_at - -
          Mouse Genome 430 2.0 Array 1443670_PM_at - -
          Mouse Genome 430 2.0 Array 1439666_at - -
          Mouse Genome 430 2.0 Array 1443670_at - -
          Mouse Genome 430 2.0 Array 1439666_PM_at - -
          Mouse Genome 430 2.0 Array 1443671_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70113_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70113_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500016502.mm.1 NM_001013022.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016502.mm.1 NM_001013022.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016502.mm.1 TC1500002053.mm.1 MTA TC
          Mouse Expression Set 430 1439666_PM_at - -
          Mouse Expression Set 430 1443671_PM_x_at - -
          Mouse Expression Set 430 1443670_at - -
          Mouse Expression Set 430 1439666_at - -
          Mouse Expression Set 430 1443670_PM_at - -
          Mouse Expression Set 430 1443671_x_at - -
          Murine Genome U74v2 Set 165925_at - -
          Murine Genome U74v2 Set 165925_PM_at - -
          Murine Genome U74v2 Set 165924_PM_at - -
          Murine Genome U74v2 Set 165924_at - -
          Affymetrix HT Mouse Genome 430B Array 1439666_at - -
          Affymetrix HT Mouse Genome 430B Array 1443670_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443671_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1443671_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1443670_at - -
          Affymetrix HT Mouse Genome 430B Array 1439666_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 438843 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1251371 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2759088 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251371 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2759088 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline