Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01299147_m1
          See other TMEM92 GE Assays ›
          Gene Symbol:
          Tmem92
          Gene Name:
          transmembrane protein 92
          Gene Aliases:
          AU016430, Tmem92-ps
          Chromosome Location:
          Chr.11: 94777217 - 94782703 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01299147_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem92
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034896.2 NP_001030068.1 1 - 2 348 94
          NM_001163172.1 NP_001156644.1 1 - 2 348 94
          GenBank mRNA AK139848.1 - 1 - 2 348 94
          BC147511.1 - 1 - 2 145 94
          BC172064.1 - 1 - 2 145 9054350 94
          BC172068.1 - 1 - 2 145 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003574.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC0700000418.rn

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P19881 AK139848.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P19881 NM_001034896.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P19881 NM_001163172.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 516842 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510476 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 817085 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 366503 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 72475 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485778 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 576818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 293496 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 766441 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 768156 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 251854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 341353 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 611581 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 48853 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142599 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 133599 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 14924 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P19881 AK139848.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P19881 NM_001034896.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P19881 NM_001163172.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02399763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28034725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28034691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28034695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28034794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28034734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28034820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28034727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28034822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28034811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28034736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28034728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28034802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28034796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28034706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28034804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28034774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09174871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09040728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09040734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09040733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09292032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09091419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28034777 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10390081 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390082 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390083 - -
          Affymetrix Mouse Gene 1.1 ST Array 10390080 - -
          Affymetrix Mouse Clariom S Array PSR1100032738.mm.1 NM_001034896.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032738.mm.1 NM_001034896.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032738.mm.1 NM_001163172.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032738.mm.1 NM_001163172.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032738.mm.1 TC1100003574.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100032740.mm.1 NM_001163172.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032740.mm.1 NM_001163172.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032740.mm.1 TC1100003574.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100032745.mm.1 NM_001034896.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032745.mm.1 NM_001034896.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032745.mm.1 NM_001163172.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032745.mm.1 NM_001163172.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032745.mm.1 TC1100003574.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100032746.mm.1 NM_001034896.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032746.mm.1 NM_001034896.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032746.mm.1 NM_001163172.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032746.mm.1 NM_001163172.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032746.mm.1 TC1100003574.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100032742.mm.1 NM_001034896.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032742.mm.1 NM_001034896.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032742.mm.1 NM_001163172.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032742.mm.1 NM_001163172.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032742.mm.1 TC1100003574.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100032739.mm.1 NM_001034896.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032739.mm.1 NM_001034896.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032739.mm.1 NM_001163172.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032739.mm.1 NM_001163172.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032739.mm.1 TC1100003574.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100032743.mm.1 NM_001034896.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032743.mm.1 NM_001034896.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032743.mm.1 NM_001163172.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032743.mm.1 NM_001163172.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032743.mm.1 TC1100003574.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 892816 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259105 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2866742 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2866742 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline