Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01299195_s1
          See other PKDREJ GE Assays ›
          Gene Symbol:
          Pkdrej
          Gene Name:
          polycystic kidney disease (polycystin) and REJ (sperm receptor for egg jelly homolog, sea urchin)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 85814676 - 85821733 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01299195_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pkdrej
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011105.2 NP_035235.2 1 6334 69
          GenBank mRNA AK029344.1 - 1 553 69

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pkdrej

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of mechanical stimulus
          regulation of acrosome reaction
          calcium ion transmembrane transport
          calcium channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30403196 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30403192 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30403193 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30403194 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P6130 NM_011105.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P6130 AK029344.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P6130 NM_011105.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P6130 AK029344.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970407 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354089 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626512 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 282634 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 124750 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175143 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P6130 NM_011105.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P6130 AK029344.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P6130 NM_011105.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P6130 AK029344.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30403218 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03089876 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175143 - -
          Whole Mouse Genome G4122A A_52_P6130 NM_011105.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P6130 AK029344.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P6130 NM_011105.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P6130 AK029344.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127352 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175143 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12120312 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12120340 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12120371 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30403193 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30403194 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30403196 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30403192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30403194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12120360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12120309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12120310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12437146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12120317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12120382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12120344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12120358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11966158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12181795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12120315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30403140 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10431227 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431228 - -
          Affymetrix HT MG-430 PM Array Plate 1422244_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10431226 - -
          Mouse Genome 430 2.0 Array 1422244_PM_at - -
          Mouse Genome 430 2.0 Array 1422244_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18766_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18766_at - -
          Affymetrix Mouse Clariom S Array PSR1500015942.mm.1 NM_011105.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015942.mm.1 NM_011105.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015942.mm.1 TC1500002008.mm.1 MTA TC
          Mouse Expression Set 430 1422244_PM_at - -
          Mouse Expression Set 430 1422244_at - -
          Mouse Genome 430A 2.0 Array 1422244_at - -
          Mouse Genome 430A 2.0 Array 1422244_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422244_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422244_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 542676 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4200088 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004200088 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2631478 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2631478 - -
          mouse-6 v1.1 (Illumina) scl46875.1.5783_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 4200088 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46875.1.5783_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4200088 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2631478 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2631478 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline