Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01299248_m1
          See other CFH GE Assays ›
          Gene Symbol:
          Cfh
          Gene Name:
          complement component factor h
          Gene Aliases:
          Mud-1, NOM, Sas-1, Sas1
          Chromosome Location:
          Chr.1: 140085855 - 140183411 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01299248_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cfh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009888.3 NP_034018.2 4 - 5 618 74
          GenBank mRNA AK052688.1 - 4 - 5 618 74
          AK087707.1 - 4 - 5 527 74
          BC066092.1 - 4 - 5 487 6842070 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003193.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cfh

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          complement activation, alternative pathway
          regulation of complement activation
          innate immune response
          complement component C3b binding
          heparin binding
          heparan sulfate proteoglycan binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100025929.mm.1 NM_009888.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025929.mm.1 NM_009888.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025929.mm.1 TC0100003193.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025932.mm.1 NM_009888.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025932.mm.1 NM_009888.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025932.mm.1 TC0100003193.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025934.mm.1 NM_009888.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025934.mm.1 NM_009888.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025934.mm.1 TC0100003193.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025930.mm.1 NM_009888.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025930.mm.1 NM_009888.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025930.mm.1 TC0100003193.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025928.mm.1 NM_009888.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025928.mm.1 NM_009888.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025928.mm.1 TC0100003193.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025927.mm.1 NM_009888.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025927.mm.1 NM_009888.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025927.mm.1 TC0100003193.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025935.mm.1 NM_009888.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025935.mm.1 NM_009888.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025935.mm.1 TC0100003193.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025931.mm.1 NM_009888.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025931.mm.1 NM_009888.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025931.mm.1 TC0100003193.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 564182 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline