Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Scube1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271472.1 | NP_001258401.1 | 8 - 9 | 1064 | 62 | |
NM_001271473.1 | NP_001258402.1 | 5 - 6 | 731 | 62 | ||
NM_022723.3 | NP_073560.2 | 7 - 8 | 974 | 62 | ||
GenBank mRNA | AF276425.1 | - | 7 - 8 | 841 | 62 | |
AK041590.1 | - | 7 - 8 | 957 | 62 | ||
BC066066.1 | - | 8 - 9 | 1022 | 6850999 | 62 | |
BC080278.1 | - | 5 - 6 | 699 | 30354974 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |