Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01299885_g1
          See other EME2 GE Assays ›
          Gene Symbol:
          Eme2
          Gene Name:
          essential meiotic endonuclease 1 homolog 2 (S. pombe)
          Gene Aliases:
          2810013J18Rik
          Chromosome Location:
          Chr.17: 24892152 - 24895087 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01299885_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eme2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163102.1 NP_001156574.1 1 - 2 268 92
          GenBank mRNA BC034637.1 - 236 92
          BC092228.1 - 1 - 2 226 92
          BY148623.1 - 1 - 2 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002799.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          DNA recombination
          cellular response to DNA damage stimulus
          biological_process
          molecular_function
          DNA binding
          nuclease activity
          endonuclease activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965757 NM_001163102.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965757 AK012738.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965757 BC092228.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 365720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 349417 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142691 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965757 NM_001163102.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965757 AK012738.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965757 BC092228.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10448759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448763 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448758 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448762 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448756 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448764 - -
          Affymetrix HT MG-430 PM Array Plate 1442853_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1460628_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1429034_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10448755 - -
          Mouse Genome 430 2.0 Array 1429034_PM_at - -
          Mouse Genome 430 2.0 Array 1429034_at - -
          Affymetrix Mouse Clariom S Array PSR1700014767.mm.1 NM_001163102.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014767.mm.1 NM_001163102.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014767.mm.1 TC1700002799.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014768.mm.1 NM_001163102.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014768.mm.1 NM_001163102.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014768.mm.1 TC1700002799.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014766.mm.1 NM_001163102.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014766.mm.1 NM_001163102.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014766.mm.1 TC1700002799.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014771.mm.1 NM_001163102.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014771.mm.1 NM_001163102.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014771.mm.1 TC1700002799.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014764.mm.1 NM_001163102.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014764.mm.1 NM_001163102.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014764.mm.1 TC1700002799.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014773.mm.1 NM_001163102.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014773.mm.1 NM_001163102.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014773.mm.1 TC1700002799.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014772.mm.1 NM_001163102.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014772.mm.1 NM_001163102.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014772.mm.1 TC1700002799.mm.1 MTA TC
          Mouse Expression Set 430 1460628_at - -
          Mouse Expression Set 430 1442853_at - -
          Mouse Expression Set 430 1442853_PM_at - -
          Mouse Expression Set 430 1429034_at - -
          Mouse Expression Set 430 1460628_PM_at - -
          Mouse Expression Set 430 1429034_PM_at - -
          Murine Genome U74v2 Set 137082_PM_at - -
          Murine Genome U74v2 Set 137082_at - -
          Affymetrix HT Mouse Genome 430B Array 1429034_at - -
          Affymetrix HT Mouse Genome 430B Array 1429034_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline