Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040187.1 | NP_001035277.1 | 2 - 3 | 694 | 152 | |
NM_199079.1 | NP_951061.1 | 2 - 3 | 513 | 152 | ||
NM_199080.2 | NP_951062.1 | 2 - 3 | 694 | 152 | ||
GenBank mRNA | AK039888.1 | - | 2 - 3 | 513 | 152 | |
AK151061.1 | - | 2 - 3 | 469 | 152 | ||
AK152749.1 | - | 2 - 3 | 469 | 152 | ||
AK152831.1 | - | 2 - 3 | 469 | 152 | ||
AK161001.1 | - | 2 - 3 | 510 | 152 | ||
AK161012.1 | - | 2 - 3 | 509 | 152 | ||
BC062910.1 | - | 2 - 3 | 499 | 6816663 | 152 | |
BC096036.1 | - | 2 - 3 | 694 | 30549388 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |