Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01300677_s1
          See other GM4371 GE Assays ›
          Gene Symbol:
          Gm4371
          Gene Name:
          eukaryotic translation initiation factor 3, subunit I pseudogene
          Gene Aliases:
          EG667874, Gm8854
          Chromosome Location:
          Chr.15: 96182800 - 96203470 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01300677_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm4371
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028311.1 2 386 93

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1995587 BY404711.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1995587 NR_028311.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1995587 BB205215.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P138298 NR_028311.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884053 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858365 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 59873 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25184 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233639 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 859481 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 768643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763811 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22793 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549325 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795467 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464950 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 60310 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 43038 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P138298 NR_028311.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1446594_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10426503 - -
          Mouse Genome 430 2.0 Array 1446594_PM_at - -
          Mouse Genome 430 2.0 Array 1446594_at - -
          Affymetrix Mouse Clariom S Array PSR1500007486.mm.1 NR_028311.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500007486.mm.1 TC1500000946.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500007487.mm.1 NR_028311.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500007487.mm.1 TC1500000946.mm.1 MTA TC
          Mouse Expression Set 430 1446594_PM_at - -
          Mouse Expression Set 430 1446594_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline