Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01300991_g1
          See other RPL10A GE Assays ›
          Gene Symbol:
          Rpl10a
          Gene Name:
          ribosomal protein L10A
          Gene Aliases:
          CsA-19, Nedd6
          Chromosome Location:
          Chr.17: 28328471 - 28331033 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01300991_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl10a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011287.2 NP_035417.2 5 - 6 627 80
          GenBank mRNA AK150743.1 - 5 - 6 515 80
          AK151172.1 - 5 - 6 516 80
          AK153863.1 - 5 - 6 513 80
          BC006039.1 - 3 - 4 844 3494193 80
          BC083346.1 - 4 - 5 493 5712034 80
          D10916.1 - 2 - 3 210 80
          U12403.1 - 5 - 6 499 80

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl10a

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          RNA binding
          structural constituent of ribosome
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31121838 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31121839 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31121832 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31121837 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31121833 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31121831 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31121835 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31121836 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2101906 BY574146.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2101906 U12403.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2101906 NM_011287.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981499 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 565608 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146213 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2101906 BY574146.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2101906 U12403.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2101906 NM_011287.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981499 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P407086 AK193763.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P407086 AK151172.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P407086 AK153863.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P407086 AK200448.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P407086 AK215262.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31121832 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146213 - -
          Whole Mouse Genome G4122A A_52_P407086 AK193763.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407086 AK151172.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407086 AK153863.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407086 AK200448.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407086 AK215262.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132545 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106415 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120812 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146213 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13161328 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13161336 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13161339 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13161340 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13161341 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13161337 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31121839 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31121832 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31121831 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31121833 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31121838 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31121837 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31121836 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31121835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31121800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13394451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13398043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13161339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13501011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13501004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13153980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13161337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13161324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13400222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13327458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13400275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13161340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31121791 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10443362 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443365 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355174 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443363 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443361 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407536 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443366 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355175 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443364 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410064 - -
          Affymetrix Mouse Gene 1.1 ST Array 10410063 - -
          Affymetrix Mouse Gene 1.1 ST Array 10443360 - -
          Affymetrix Mouse Gene 1.1 ST Array 10355173 - -
          Affymetrix Mouse Gene 1.1 ST Array 10407535 - -
          Mouse Genome 430 2.0 Array 1431177_PM_a_at - -
          Mouse Genome 430 2.0 Array 1431177_a_at - -
          Affymetrix Mouse Clariom S Array PSR1700004153.mm.1 TC1700000477.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700004151.mm.1 TC1700000477.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700004159.mm.1 NM_011287.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004159.mm.1 NM_011287.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004159.mm.1 TC1700000477.mm.1 MTA TC
          Mouse Expression Set 430 1431177_a_at - -
          Mouse Expression Set 430 1431177_PM_a_at - -
          Murine Genome U74v2 Set 161327_f_at - -
          Murine Genome U74v2 Set 100711_at - -
          Murine Genome U74v2 Set 161327_PM_f_at - -
          Murine Genome U74v2 Set 100711_PM_at - -
          Mouse Genome 430A 2.0 Array 1431177_a_at - -
          Mouse Genome 430A 2.0 Array 1431177_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 897428 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 104060020 - -
          Illumina mouse-6 v1.0 expression beadchip 4850066 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004850066 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2746168 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2746168 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225260 - -
          mouse-6 v1.1 (Illumina) scl019896.4_30-S - -
          mouse-6 v1.1 (Illumina) GI_6755349-S - -
          Illumina Sentrix mouse-6.v1 bead array 104060020 - -
          Illumina Sentrix mouse-6.v1 bead array 4850066 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl019896.4_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4850066 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104060020 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2902115 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl019896.4_30-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline