| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10448907 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448898 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448882 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448887 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448908 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448894 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10442761 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448901 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448886 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448906 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448910 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448896 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448891 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448880 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448890 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448909 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448897 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448884 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448903 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448883 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448893 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448888 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448904 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448879 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448899 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448905 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448885 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448911 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448902 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448889 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448895 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448881 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1427509_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10448878 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10442752 |
- |
- |
| Mouse Genome 430 2.0 Array |
1427509_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1427509_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1700014999.mm.1 |
NM_001163270.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014999.mm.1 |
NM_001163270.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014999.mm.1 |
TC1700001656.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700014995.mm.1 |
NM_001163270.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014995.mm.1 |
NM_001163270.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014995.mm.1 |
TC1700001656.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700015000.mm.1 |
NM_001163270.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015000.mm.1 |
NM_001163270.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015000.mm.1 |
TC1700001656.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700015001.mm.1 |
NM_001163270.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015001.mm.1 |
NM_001163270.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015001.mm.1 |
TC1700001656.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700014998.mm.1 |
NM_001163270.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014998.mm.1 |
NM_001163270.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014998.mm.1 |
TC1700001656.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1427509_PM_at |
- |
- |
| Mouse Expression Set 430 |
1427509_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1427509_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1427509_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1427509_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1427509_PM_at |
- |
- |