Search
Search

Wdr90| Gene Symbol: Wdr90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163766.1 | NP_001157238.1 | 12 - 13 | 1735 | 61 | |
| GenBank mRNA | AK014328.1 | - | 8 - 9 | 836 | 61 | |
| AK036541.1 | - | 5 - 6 | 2764 | 61 | ||
| AK129465.1 | - | 11 - 12 | 1650 | 61 | ||
| AK154827.1 | - | 10 - 11 | 1902 | 61 | ||
| AK170906.1 | - | 12 - 13 | 1695 | 61 | ||
Set Membership: |
|