Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01301287_m1
          See other FBXL16 GE Assays ›
          Gene Symbol:
          Fbxl16
          Gene Name:
          F-box and leucine-rich repeat protein 16
          Gene Aliases:
          BC042620, Scirr1
          Chromosome Location:
          Chr.17: 25809050 - 25821265 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01301287_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fbxl16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164225.1 NP_001157697.1 2 - 3 946 62
          GenBank mRNA AK205343.1 - 1 - 2 187 62
          BC042620.1 - 1 - 2 655 62
          BC132383.1 - 1 - 2 742 40130660 62
          BC137657.1 - 1 - 2 742 8860676 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000389.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P49457 BC042620.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P49457 NM_001164225.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P49457 BC042620.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P49457 NM_001164225.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155990 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P49457 BC042620.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P49457 NM_001164225.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P49457 BC042620.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P49457 NM_001164225.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155990 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08743675 - -
          Whole Mouse Genome G4122A A_52_P49457 BC042620.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P49457 NM_001164225.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P49457 BC042620.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P49457 NM_001164225.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101535 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13286795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13132843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106418 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106380 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442889 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442888 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442892 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442891 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442890 - -
          Affymetrix HT MG-430 PM Array Plate 1440286_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442887 - -
          Mouse Genome 430 2.0 Array 1440286_PM_at - -
          Mouse Genome 430 2.0 Array 1440286_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 214931_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 214931_at - -
          Affymetrix Mouse Clariom S Array PSR1700003265.mm.1 NM_001164225.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003265.mm.1 NM_001164225.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003265.mm.1 TC1700000389.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003264.mm.1 NM_001164225.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003264.mm.1 NM_001164225.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003264.mm.1 TC1700000389.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003263.mm.1 TC1700000389.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003262.mm.1 NM_001164225.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003262.mm.1 NM_001164225.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003262.mm.1 TC1700000389.mm.1 MTA TC
          Mouse Expression Set 430 1440286_at - -
          Mouse Expression Set 430 1440286_PM_at - -
          Murine Genome U74v2 Set 139278_at - -
          Murine Genome U74v2 Set 139278_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440286_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440286_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 463583 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2653194 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2653194 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline