Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01302368_m1
          See other TRAPPC6A GE Assays ›
          Gene Symbol:
          Trappc6a
          Gene Name:
          trafficking protein particle complex 6A
          Gene Aliases:
          1810073E21Rik, 4930519D19Rik, AI480686, TRS33, mhyp
          Chromosome Location:
          Chr.7: 19507406 - 19516145 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01302368_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trappc6a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025960.3 NP_080236.2 1 - 2 100 158
          GenBank mRNA AK007965.1 - 1 - 2 99 158
          AK008022.1 - 1 - 2 100 158
          AK008462.2 - 1 - 2 99 158
          AK088832.1 - 1 - 2 99 158
          BC037154.1 - 1 - 2 137 2811950 158
          BC106114.1 - 1 - 2 114 4168174 158

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000303.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trappc6a

          Gene Ontology Categories:

          Function(s)

          transport
          vesicle-mediated transport

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24993672 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24993772 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24993673 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24993775 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24993748 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24993776 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24993674 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24993770 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24993747 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106250 BC106114.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106250 BC020128.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106250 BC037154.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106250 NM_025960.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106250 AK007965.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628429 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 138808 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 581321 AF105222.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 581321 BC046752.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 581321 NM_019675.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 24193 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 1402 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 9177 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104674 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106250 BC106114.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106250 BC020128.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106250 BC037154.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106250 NM_025960.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106250 AK007965.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24993679 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01484161 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104674 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06135354 - -
          Whole Mouse Genome G4122A A_51_P485683 BC106114.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P485683 AK088832.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P485683 BC020128.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P485683 BC037154.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P485683 NM_025960.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140038 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138451 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104674 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06601463 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06601467 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06601469 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24993772 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24993776 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24993672 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24993747 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24993674 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24993748 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24993775 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24993770 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24993673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24993712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06295586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06295577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06027122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06564910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06658124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06442589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06174579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06601473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06295583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06449800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06295574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05944118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06449798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06442584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06197471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06027127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06389153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06197472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24993774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06402238 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550722 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550720 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550717 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550721 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550718 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550716 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550719 - -
          Affymetrix HT MG-430 PM Array Plate 1417034_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1460054_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10550715 - -
          Mouse Genome 430 2.0 Array 1460054_at - -
          Mouse Genome 430 2.0 Array 1417034_at - -
          Mouse Genome 430 2.0 Array 1460054_PM_at - -
          Mouse Genome 430 2.0 Array 1417034_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67091_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67091_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700002423.mm.1 TC0700000303.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002423.mm.1 NM_025960.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002421.mm.1 TC0700000303.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002421.mm.1 NM_025960.4 Refseq
          Mouse Expression Set 430 1417034_at - -
          Mouse Expression Set 430 1417034_PM_at - -
          Mouse Expression Set 430 1460054_PM_at - -
          Mouse Expression Set 430 1460054_at - -
          Mouse Genome 430A 2.0 Array 1417034_PM_at - -
          Mouse Genome 430A 2.0 Array 1417034_at - -
          Affymetrix HT Mouse Genome 430A Array 1417034_at - -
          Affymetrix HT Mouse Genome 430A Array 1417034_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1460054_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1460054_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 931594 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 70717 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000070717 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2643057 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2643057 - -
          mouse-6 v1.1 (Illumina) scl067091.5_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 70717 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067091.5_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 70717 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2643057 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2643057 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067091.5_7-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline