Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01302477_g1
          See other HCFC1R1 GE Assays ›
          Gene Symbol:
          Hcfc1r1
          Gene Name:
          host cell factor C1 regulator 1 (XPO1-dependent)
          Gene Aliases:
          HPIP
          Chromosome Location:
          Chr.17: 23673608 - 23675524 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01302477_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hcfc1r1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181821.1 NP_861542.1 1 - 2 464 60
          GenBank mRNA AK003865.1 - 1 - 2 117 60
          AK013438.1 - 1 - 2 464 60
          BC079865.1 - 1 - 2 104 6818222 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000315.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hcfc1r1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31092954 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31092955 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P582112 AK013438.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P582112 NM_181821.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P582112 AK003865.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P582112 BC079865.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P582112 AK013438.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P582112 NM_181821.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P582112 AK003865.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P582112 BC079865.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586909 AK047071.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586909 AK075689.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586909 AK077034.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586909 AK147468.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586909 AK147470.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586909 AK147506.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586909 AK147696.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586909 BC023453.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586909 BC034081.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586909 BC039756.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586909 NM_001039086.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586909 NM_001039087.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586909 NM_054050.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 160175 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119820 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P582112 AK013438.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P582112 NM_181821.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P582112 AK003865.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P582112 BC079865.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P582112 AK013438.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P582112 NM_181821.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P582112 AK003865.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P582112 BC079865.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092949 - -
          Whole Mouse Genome G4122A A_52_P582112 AK013438.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P582112 NM_181821.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P582112 AK003865.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P582112 BC079865.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P582112 AK013438.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P582112 NM_181821.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P582112 AK003865.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P582112 BC079865.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100127 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13383006 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13214728 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13214693 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13214736 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31092955 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31092954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13368437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13360858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13323326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13405511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13360859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13360854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13405512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092944 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442323 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442325 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442324 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448305 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448306 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442322 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442326 - -
          Affymetrix HT MG-430 PM Array Plate 1428405_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1428406_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442321 - -
          Affymetrix Mouse Gene 1.1 ST Array 10448292 - -
          Mouse Genome 430 2.0 Array 1428406_s_at - -
          Mouse Genome 430 2.0 Array 1428405_at - -
          Mouse Genome 430 2.0 Array 1428406_PM_s_at - -
          Mouse Genome 430 2.0 Array 1428405_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 353502_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 353502_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700002291.mm.1 NM_181821.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002291.mm.1 NM_181821.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002291.mm.1 TC1700000315.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002298.mm.1 NM_181821.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002298.mm.1 NM_181821.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002298.mm.1 TC1700000315.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002297.mm.1 NM_181821.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002297.mm.1 NM_181821.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002297.mm.1 TC1700000315.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002290.mm.1 NM_181821.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002290.mm.1 NM_181821.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002290.mm.1 TC1700000315.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002293.mm.1 NM_181821.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002293.mm.1 NM_181821.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002293.mm.1 TC1700000315.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002296.mm.1 NM_181821.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002296.mm.1 NM_181821.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002296.mm.1 TC1700000315.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002295.mm.1 NM_181821.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002295.mm.1 NM_181821.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002295.mm.1 TC1700000315.mm.1 MTA TC
          Mouse Expression Set 430 1428406_PM_s_at - -
          Mouse Expression Set 430 1428406_s_at - -
          Mouse Expression Set 430 1428405_at - -
          Mouse Expression Set 430 1428405_PM_at - -
          Murine Genome U74v2 Set 94266_PM_at - -
          Murine Genome U74v2 Set 94266_at - -
          Mouse Genome 430A 2.0 Array 1428405_PM_at - -
          Mouse Genome 430A 2.0 Array 1428405_at - -
          Mouse Genome 430A 2.0 Array 1428406_s_at - -
          Mouse Genome 430A 2.0 Array 1428406_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1428405_at - -
          Affymetrix HT Mouse Genome 430A Array 1428406_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1428405_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428406_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 666974 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6350500 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006350500 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2723083 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2723083 - -
          mouse-6 v1.1 (Illumina) scl51033.3_356-S - -
          Illumina Sentrix mouse-6.v1 bead array 6350500 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl51033.3_356-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6350500 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2723083 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2723083 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline