Search
Search

0610007P14Rik| Gene Symbol: 0610007P14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021446.2 | NP_067421.1 | 3 - 4 | 319 | 65 | |
| GenBank mRNA | AF270646.1 | - | 3 - 4 | 318 | 65 | |
| AK004480.1 | - | 3 - 4 | 341 | 65 | ||
| AK152230.1 | - | 3 - 4 | 297 | 65 | ||
| BC004591.1 | - | 3 - 4 | 305 | 3587693 | 65 | |
Set Membership: |
|