Search
Search
| Gene Symbol: Wsb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042565.3 | NP_001036030.1 | 2 - 3 | 1162 | 67 | |
| NM_019653.3 | NP_062627.3 | 7 - 8 | 1308 | 67 | ||
| GenBank mRNA | AF033186.1 | - | 7 - 8 | 1005 | 67 | |
| AK019206.2 | - | 7 - 8 | 1285 | 67 | ||
| AK048511.1 | - | 2 - 3 | 1162 | 67 | ||
| AK145974.1 | - | 7 - 8 | 1287 | 67 | ||
| AK146752.1 | - | 7 - 8 | 1283 | 67 | ||
| AK150394.1 | - | 7 - 8 | 1283 | 67 | ||
| AK150600.1 | - | 7 - 8 | 1283 | 67 | ||
| AK152545.1 | - | 7 - 8 | 1286 | 67 | ||
| AK159905.1 | - | 7 - 8 | 1283 | 67 | ||
| AK167174.1 | - | 7 - 8 | 1285 | 67 | ||
| AK167394.1 | - | 7 - 8 | 1286 | 67 | ||
| AK167942.1 | - | 7 - 8 | 1283 | 67 | ||
| AK168468.1 | - | 7 - 8 | 2706 | 67 | ||
| AK168565.1 | - | 7 - 8 | 2705 | 67 | ||
| AK169058.1 | - | 7 - 8 | 1283 | 67 | ||
| BC019601.1 | - | 2 - 3 | 378 | 67 | ||
| BC053391.1 | - | 7 - 8 | 1059 | 30073116 | 67 | |
Set Membership: |
|