Search
Search

Hnrnpa1| Gene Symbol: Hnrnpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104427.1 | 10 - 11 | 1170 | 99 | ||
| GenBank mRNA | AK136113.1 | - | 10 - 11 | 1140 | 99 | |
| BC052723.1 | - | 12 - 13 | 2968 | 99 | ||
| CJ235774.1 | - | 1 - 2 | 311 | 99 | ||
Set Membership: |
|