| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700016566.mm.1 |
NM_001081160.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700016566.mm.1 |
NM_001081160.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700016566.mm.1 |
TC1700001794.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700016568.mm.1 |
NM_001081160.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700016568.mm.1 |
NM_001081160.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700016568.mm.1 |
TC1700001794.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700016567.mm.1 |
NM_001081160.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700016567.mm.1 |
NM_001081160.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700016567.mm.1 |
TC1700001794.mm.1 |
MTA TC |