| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500015773.mm.1 |
NM_001163145.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015773.mm.1 |
NM_001163145.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015773.mm.1 |
TC1500001987.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015772.mm.1 |
NM_001163145.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015772.mm.1 |
NM_001163145.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015772.mm.1 |
TC1500001987.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015774.mm.1 |
NM_001163145.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015774.mm.1 |
NM_001163145.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015774.mm.1 |
TC1500001987.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015775.mm.1 |
NM_001163145.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015775.mm.1 |
NM_001163145.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015775.mm.1 |
TC1500001987.mm.1 |
MTA TC |