Search
Search

2410089E03Rik| Gene Symbol: 2410089E03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162906.1 | NP_001156378.1 | 3 - 4 | 600 | 106 | |
| GenBank mRNA | AK028897.1 | - | 3 - 4 | 601 | 106 | |
Set Membership: |
|