| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27953993 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27953994 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27953992 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P549927 |
AK160515.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P549927 |
BC044897.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P549927 |
BC055024.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P549927 |
AK135352.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P549927 |
AK153246.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P549927 |
AK160515.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P549927 |
BC044897.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P549927 |
BC055024.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P549927 |
AK135352.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P549927 |
AK153246.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
396124 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
AK010854.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
AK014094.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
AK018849.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
AK159788.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
AK166904.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
AK166995.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
AK167396.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
AK167633.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
AK167935.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
AK168670.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
AK195168.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
AK202921.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
AK209718.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
AK211423.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
AK216587.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
BC005559.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
BC057973.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
BC083119.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545160 |
NM_025607.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
833383 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
364801 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
585244 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
157327 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
52499 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
246093 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
478644 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
432118 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
90937 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
19961 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P116328 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P104059 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P549927 |
AK160515.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P549927 |
BC044897.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P549927 |
BC055024.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P549927 |
AK135352.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P549927 |
AK153246.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P549927 |
AK160515.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P549927 |
BC044897.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P549927 |
BC055024.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P549927 |
AK135352.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P549927 |
AK153246.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P116328 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07413284 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P104059 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P169564 |
AK135352.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P169564 |
AK078548.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P169564 |
NM_027472.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P460119 |
NM_027472.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P549929 |
BC044897.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P549929 |
BC055024.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P549929 |
AK135352.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P549929 |
AK153246.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P549929 |
AK078548.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P549927 |
AK160515.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P549927 |
BC044897.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P549927 |
BC055024.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P549927 |
AK135352.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P549927 |
AK153246.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P549927 |
AK160515.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P549927 |
BC044897.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P549927 |
BC055024.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P549927 |
AK135352.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P549927 |
AK153246.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P104059 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P116328 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09088599 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09234199 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09573291 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09645874 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09573296 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09285993 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09716871 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27953994 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27953992 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27953993 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953943 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953947 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953970 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953991 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953959 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953969 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954036 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953952 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953980 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954015 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953967 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953957 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953920 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953940 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953927 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954027 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953979 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954034 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954017 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953958 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953910 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953929 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954018 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953935 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953988 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953993 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953914 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954013 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954010 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953976 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953956 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953999 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953937 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953938 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953977 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954021 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953919 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954037 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953971 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954009 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953982 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953964 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953912 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953974 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954031 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953985 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954020 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954026 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953954 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954028 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953955 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954005 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954014 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954032 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954003 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953962 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27954023 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953933 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953939 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953904 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953931 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953966 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953908 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953949 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27953995 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953971 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953999 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953979 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953981 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27954001 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953970 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953998 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953977 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953962 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953960 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953993 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953963 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09525374 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09509672 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953969 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09329029 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953997 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09716870 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09034582 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953989 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953995 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953988 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953982 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09060590 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953974 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27953966 |
- |
- |