Search
Search

Trim25| Gene Symbol: Trim25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009546.2 | NP_033572.2 | 6 - 7 | 1267 | 117 | |
| GenBank mRNA | AK154533.1 | - | 6 - 7 | 1260 | 117 | |
| AK169562.1 | - | 6 - 7 | 1268 | 117 | ||
| AK171744.1 | - | 5 - 6 | 571 | 117 | ||
| D63902.1 | - | 6 - 7 | 1190 | 117 | ||
Set Membership: |
|