Search
Search

Acaca| Gene Symbol: Acaca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133360.2 | NP_579938.2 | 27 - 28 | 3600 | 62 | |
| GenBank mRNA | AF289714.1 | - | 1 - 2 | 74 | 62 | |
| AY451393.1 | - | 27 - 28 | 3574 | 62 | ||
| BC042725.1 | - | 8 - 9 | 933 | 62 | ||
Set Membership: |
|