| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300007606.mm.1 |
NR_045083.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007606.mm.1 |
NR_045083.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007606.mm.1 |
NR_045084.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007606.mm.1 |
NR_045084.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007606.mm.1 |
TC1300001194.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300007607.mm.1 |
NR_045084.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007607.mm.1 |
NR_045084.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007607.mm.1 |
TC1300001194.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300007608.mm.1 |
NR_045084.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007608.mm.1 |
NR_045084.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007608.mm.1 |
TC1300001194.mm.1 |
MTA TC |