Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01305725_g1
          See other PDIA2 GE Assays ›
          Gene Symbol:
          Pdia2
          Gene Name:
          protein disulfide isomerase associated 2
          Gene Aliases:
          1810041F13Rik, AI661267, Pdip, Pdipl
          Chromosome Location:
          Chr.17: 26195999 - 26199132 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01305725_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pdia2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081070.1 NP_001074539.1 10 - 11 1595 82
          GenBank mRNA AK007743.1 - 10 - 11 1544 82
          BC116671.1 - 10 - 11 1561 40107256 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002806.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          response to endoplasmic reticulum stress
          cell redox homeostasis
          regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          protein disulfide isomerase activity
          steroid binding
          lipid binding
          isomerase activity
          peptidyl-proline 4-dioxygenase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133057 NM_001081070.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133057 AK007743.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2131964 NM_001081070.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2131964 AK007743.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2131964 BC116671.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 699213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 33517 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 576096 AK020596.1 genbank_mrna_mouse
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131964 NM_001081070.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131964 AK007743.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131964 BC116671.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133057 NM_001081070.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133057 AK007743.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P227984 NM_001081070.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P227984 AK007743.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P227984 BC116671.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31108929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31108854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31108865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13157867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31108855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31108864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31108877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31108857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31108870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31108879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13157834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31108867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31108882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31108873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31108878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13157843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31108875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31108880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31108883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31108868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31108872 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10449255 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449254 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449249 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449251 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449256 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449253 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449257 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449246 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449248 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449250 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449247 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449252 - -
          Affymetrix HT MG-430 PM Array Plate 1428952_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10449245 - -
          Mouse Genome 430 2.0 Array 1428952_at - -
          Mouse Genome 430 2.0 Array 1428952_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69191_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69191_at - -
          Affymetrix Mouse Clariom S Array PSR1700015705.mm.1 NM_001081070.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015705.mm.1 NM_001081070.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015705.mm.1 TC1700002806.mm.1 MTA TC
          Mouse Expression Set 430 1428952_PM_at - -
          Mouse Expression Set 430 1428952_at - -
          Murine Genome U74v2 Set 170234_r_at - -
          Murine Genome U74v2 Set 170234_PM_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1428952_at - -
          Affymetrix HT Mouse Genome 430B Array 1428952_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 311434 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215125 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215125 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline