Search
Search

Ppil1| Gene Symbol: Ppil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026845.4 | NP_081121.1 | 1 - 2 | 102 | 58 | |
| GenBank mRNA | AK004331.1 | - | 1 - 2 | 103 | 58 | |
| AK150643.1 | - | 1 - 2 | 79 | 58 | ||
| AK151572.1 | - | 1 - 2 | 79 | 58 | ||
| AK156809.1 | - | 1 - 2 | 103 | 58 | ||
| BC037131.1 | - | 1 - 2 | 82 | 58 | ||
| BC051386.1 | - | 1 - 2 | 69 | 58 | ||
| BC058369.1 | - | 1 - 2 | 61 | 5715252 | 58 | |
| BC147647.1 | - | 1 - 2 | 187 | 9056637 | 58 | |
| BC147649.1 | - | 1 - 2 | 187 | 9056639 | 58 | |
Set Membership: |
|