Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01306319_m1
          See other WBP2NL GE Assays ›
          Gene Symbol:
          Wbp2nl
          Gene Name:
          WBP2 N-terminal like
          Gene Aliases:
          4930521I23Rik, PAWP
          Chromosome Location:
          Chr.15: 82298984 - 82314558 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01306319_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wbp2nl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029066.1 NP_083342.1 5 - 6 539 105
          GenBank mRNA AK015863.1 - 5 - 6 570 105
          BC119520.2 - 5 - 6 539 40055925 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000748.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          egg activation
          female pronucleus assembly
          male pronucleus assembly
          WW domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161485 NM_029066.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161485 AK015863.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161485 BC119520.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161485 NM_029066.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161485 AK015863.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161485 BC119520.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 173213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510729 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 571705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418841 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478910 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744504 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 689403 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 700399 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 23388 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 152689 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164354 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110979 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161485 NM_029066.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161485 AK015863.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161485 BC119520.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161485 NM_029066.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161485 AK015863.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161485 BC119520.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03083912 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164354 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110979 - -
          Whole Mouse Genome G4122A A_51_P161485 NM_029066.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161485 AK015863.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161485 BC119520.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161485 NM_029066.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161485 AK015863.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161485 BC119520.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164354 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30382765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11951294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382693 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11941600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12195512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12084129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11870730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30382651 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10425744 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425745 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425747 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425746 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425748 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425743 - -
          Affymetrix HT MG-430 PM Array Plate 1431578_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10425741 - -
          Mouse Genome 430 2.0 Array 1431578_at - -
          Mouse Genome 430 2.0 Array 1431578_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74716_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74716_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500005905.mm.1 NM_029066.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005905.mm.1 NM_029066.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005905.mm.1 TC1500000748.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005907.mm.1 NM_029066.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005907.mm.1 NM_029066.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005907.mm.1 TC1500000748.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005904.mm.1 NM_029066.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005904.mm.1 NM_029066.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005904.mm.1 TC1500000748.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005902.mm.1 NM_029066.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005902.mm.1 NM_029066.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005902.mm.1 TC1500000748.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005906.mm.1 NM_029066.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005906.mm.1 NM_029066.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005906.mm.1 TC1500000748.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005903.mm.1 NM_029066.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005903.mm.1 NM_029066.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005903.mm.1 TC1500000748.mm.1 MTA TC
          Mouse Expression Set 430 1431578_at - -
          Mouse Expression Set 430 1431578_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431578_at - -
          Affymetrix HT Mouse Genome 430B Array 1431578_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 687842 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_3004065 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3004065 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline