Search
Search

Cwc25| Gene Symbol: Cwc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026186.4 | NP_080462.2 | 8 - 9 | 1143 | 57 | |
| GenBank mRNA | AK004983.2 | - | 8 - 9 | 1087 | 57 | |
| AK017849.1 | - | 8 - 9 | 1083 | 57 | ||
| AK035158.1 | - | 8 - 9 | 1071 | 57 | ||
| BC053447.1 | - | 1 - 2 | 297 | 57 | ||
Set Membership: |
|