Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01308267_m1
          See other LYRM9 GE Assays ›
          Gene Symbol:
          Lyrm9
          Gene Name:
          LYR motif containing 9
          Gene Aliases:
          1810012P15Rik
          Chromosome Location:
          Chr.11: 78826595 - 78843899 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01308267_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lyrm9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001076681.2 NP_001070149.1 2 - 3 179 71
          GenBank mRNA AK007467.1 - 2 - 3 193 71
          AK009070.1 - 2 - 3 162 71
          AK144380.1 - 2 - 3 180 71
          BC130224.1 - 2 - 3 170 40141726 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001169.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27945542 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27945544 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27945545 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27945543 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27945541 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P393120 BC031726.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P393120 NM_001076681.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P393120 BC031726.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P393120 NM_001076681.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 139918 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 571787 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813038 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413459 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 261775 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736153 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923925 AF139659.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923925 AK078405.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923925 AK163256.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923925 BC048689.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923925 BC080739.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923925 NM_019716.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 859350 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 819517 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753189 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 51594 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138970 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P393120 BC031726.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P393120 NM_001076681.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P393120 BC031726.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P393120 NM_001076681.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138970 - -
          Whole Mouse Genome G4122A A_52_P540462 BC130224.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P540462 NM_001076681.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P540462 AK144380.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P134146 BC130224.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P134146 NM_001076681.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P134146 AK007467.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P134146 AK009070.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P134146 AK144380.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P393120 BC031726.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P393120 NM_001076681.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P393120 BC031726.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P393120 NM_001076681.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100129 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138970 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27945542 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27945545 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27945543 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27945544 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27945541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09522890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09297672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09297670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08994957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09399142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09348926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09042493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09355296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08994958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09155457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09574529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09155448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09155463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09616862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945495 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10379226 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379227 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379224 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379225 - -
          Affymetrix HT MG-430 PM Array Plate 1458028_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440234_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1436548_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10379223 - -
          Mouse Genome 430 2.0 Array 1436548_PM_at - -
          Mouse Genome 430 2.0 Array 1436548_at - -
          Mouse Genome 430 2.0 Array 1458028_PM_at - -
          Mouse Genome 430 2.0 Array 1440234_PM_at - -
          Mouse Genome 430 2.0 Array 1458028_at - -
          Mouse Genome 430 2.0 Array 1440234_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66274_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66274_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100010902.mm.1 NM_001076681.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010902.mm.1 NM_001076681.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010902.mm.1 TC1100001169.mm.1 MTA TC
          Mouse Expression Set 430 1436548_at - -
          Mouse Expression Set 430 1458028_at - -
          Mouse Expression Set 430 1436548_PM_at - -
          Mouse Expression Set 430 1440234_at - -
          Mouse Expression Set 430 1458028_PM_at - -
          Mouse Expression Set 430 1440234_PM_at - -
          Murine Genome U74v2 Set 130033_PM_at - -
          Murine Genome U74v2 Set 130033_at - -
          Affymetrix HT Mouse Genome 430B Array 1440234_at - -
          Affymetrix HT Mouse Genome 430B Array 1440234_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1458028_at - -
          Affymetrix HT Mouse Genome 430B Array 1458028_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436548_at - -
          Affymetrix HT Mouse Genome 430B Array 1436548_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 850575 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 940164 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1229428 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229428 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline