Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01308573_g1
          See other GM13119 GE Assays ›
          Gene Symbol:
          Gm13119
          Gene Name:
          predicted gene 13119
          Gene Aliases:
          OTTMUSG00000010537
          Chromosome Location:
          Chr.4: 144357964 - 144364419 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01308573_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm13119
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034101.2 NP_001029273.2 3 - 4 923 75
          GenBank mRNA BC100406.1 - 2 - 3 923 30727972 75

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gm13119

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959148 BC100406.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959148 NM_001034101.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959146 BC100406.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959146 NM_001034101.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959146 BC100406.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959146 NM_001034101.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959148 BC100406.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959148 NM_001034101.2 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378210 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10510071 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510075 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510070 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510073 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510069 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510115 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510116 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510114 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510079 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510078 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510074 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510077 - -
          Affymetrix Mouse Gene 1.1 ST Array 10510112 - -
          Affymetrix Mouse Gene 1.1 ST Array 10510076 - -
          Affymetrix Mouse Gene 1.1 ST Array 10510072 - -
          Affymetrix Mouse Gene 1.1 ST Array 10510068 - -
          Affymetrix Mouse Clariom S Array PSR0400015195.mm.1 NM_001034101.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015195.mm.1 NM_001034101.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015195.mm.1 TC0400001848.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251942 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2938993 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2938993 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0