Search
Search
| Gene Symbol: Emb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010330.4 | NP_034460.3 | 6 - 7 | 1160 | 123 | |
| GenBank mRNA | AK079570.1 | - | 6 - 7 | 1102 | 123 | |
| AK088480.1 | - | 6 - 7 | 988 | 123 | ||
| AK138271.1 | - | 6 - 7 | 1109 | 123 | ||
| AK148449.1 | - | 6 - 7 | 988 | 123 | ||
| AK148540.1 | - | 6 - 7 | 988 | 123 | ||
| AK150625.1 | - | 6 - 7 | 989 | 123 | ||
| AK161679.1 | - | 6 - 7 | 988 | 123 | ||
| AK166602.1 | - | 6 - 7 | 988 | 123 | ||
| AK167997.1 | - | 6 - 7 | 1020 | 123 | ||
| BC014858.1 | - | 6 - 7 | 979 | 4500922 | 123 | |
Set Membership: |
|