Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01308734_s1
          See other OLFR287 GE Assays ›
          Gene Symbol:
          Olfr287
          Gene Name:
          olfactory receptor 287
          Gene Aliases:
          MOR286-1
          Chromosome Location:
          Chr.15: 98204566 - 98221056 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01308734_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr287
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011780.1 NP_001011780.1 3 1363 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 488989 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 194244 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 607931 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 368466 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 27105 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 107810 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 798423 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174553 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09547537 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09547533 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09547536 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09547536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30473758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32943604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32943605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12312687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12320538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30473737 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500017596.mm.1 NM_001011780.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017596.mm.1 NM_001011780.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017596.mm.1 TC1500002353.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500017599.mm.1 NM_001011780.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017599.mm.1 NM_001011780.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017599.mm.1 TC1500002353.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500017606.mm.1 NM_001011780.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017606.mm.1 NM_001011780.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017606.mm.1 TC1500002353.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500017607.mm.1 NM_001011780.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017607.mm.1 NM_001011780.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017607.mm.1 TC1500002353.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500017600.mm.1 NM_001011780.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017600.mm.1 NM_001011780.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017600.mm.1 TC1500002353.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500017604.mm.1 NM_001011780.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017604.mm.1 NM_001011780.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017604.mm.1 TC1500002353.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500017598.mm.1 NM_001011780.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017598.mm.1 NM_001011780.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017598.mm.1 TC1500002353.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500017597.mm.1 NM_001011780.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017597.mm.1 NM_001011780.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017597.mm.1 TC1500002353.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500017603.mm.1 NM_001011780.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017603.mm.1 NM_001011780.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017603.mm.1 TC1500002353.mm.1 MTA TC
          Mouse Expression Set 430 1457225_at - -
          Mouse Expression Set 430 1457225_PM_at - -
          Murine Genome U74v2 Set 136943_s_at - -
          Murine Genome U74v2 Set 136943_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 729971 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0