Search
Search

Pnpla5| Gene Symbol: Pnpla5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029427.1 | NP_083703.1 | 5 - 6 | 827 | 83 | |
| GenBank mRNA | AK014771.1 | - | 5 - 6 | 828 | 83 | |
| BC109360.2 | - | 5 - 6 | 827 | 40058461 | 83 | |
| BC109361.2 | - | 5 - 6 | 827 | 40058463 | 83 | |
Set Membership: |
|