Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01309093_m1
          See other 4930539E08RIK GE Assays ›
          Gene Symbol:
          4930539E08Rik
          Gene Name:
          RIKEN cDNA 4930539E08 gene
          Gene Aliases:
          9130014G02
          Chromosome Location:
          Chr.17: 28896395 - 28915336 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01309093_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930539E08Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172450.3 NP_766038.1 10 - 11 1798 83
          GenBank mRNA AK033585.1 - 10 - 11 1799 83
          BC117930.2 - 10 - 11 1794 40097681 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001771.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 4930539E08Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P179480 AK033585.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P179480 AK033585.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 602352 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 1833 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 102759 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 936353 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 703338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 115215 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692273 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 220422 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727954 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 862881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 206777 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 65107 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 131356 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157217 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106870 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P179480 AK033585.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P179480 AK033585.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31125007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124999 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03301689 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106870 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157217 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08751965 - -
          Whole Mouse Genome G4122A A_51_P179480 AK033585.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P179480 AK033585.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P198802 AK019726.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117120 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106870 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157217 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13216649 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13262141 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13216644 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13216647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31125011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13166389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13244217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13339484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13191621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13216647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13216651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13487668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13191622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13199549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31125014 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10449518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449521 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449522 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449513 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449514 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449517 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449511 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449512 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449519 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449515 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449520 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449516 - -
          Affymetrix HT MG-430 PM Array Plate 1437918_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10449510 - -
          Mouse Genome 430 2.0 Array 1437918_PM_at - -
          Mouse Genome 430 2.0 Array 1437918_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 207819_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 207819_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700016326.mm.1 NM_172450.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016326.mm.1 NM_172450.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016326.mm.1 TC1700001771.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700016328.mm.1 NM_172450.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016328.mm.1 NM_172450.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016328.mm.1 TC1700001771.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700016325.mm.1 NM_172450.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016325.mm.1 NM_172450.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016325.mm.1 TC1700001771.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700016327.mm.1 NM_172450.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016327.mm.1 NM_172450.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016327.mm.1 TC1700001771.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700016329.mm.1 NM_172450.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016329.mm.1 NM_172450.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016329.mm.1 TC1700001771.mm.1 MTA TC
          Mouse Expression Set 430 1437918_at - -
          Mouse Expression Set 430 1437918_PM_at - -
          Murine Genome U74v2 Set 166377_i_at - -
          Murine Genome U74v2 Set 166377_PM_i_at - -
          Affymetrix HT Mouse Genome 430B Array 1437918_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1437918_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 632987 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6380324 - -
          Illumina mouse-6 v1.0 expression beadchip 6380324 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006380324 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1238069 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238069 - -
          mouse-6 v1.1 (Illumina) scl50111.12.1_239-S - -
          Illumina Sentrix mouse-6.v1 bead array 6380324 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50111.12.1_239-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6380324 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1238069 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238069 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50111.12.1_239-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline