Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01309652_m1
          See other FNDC8 GE Assays ›
          Gene Symbol:
          Fndc8
          Gene Name:
          fibronectin type III domain containing 8
          Gene Aliases:
          4930466G16Rik
          Chromosome Location:
          Chr.11: 82892145 - 82900737 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01309652_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fndc8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030224.1 NP_084500.1 3 - 4 865 66
          GenBank mRNA AK019639.1 - 3 - 4 887 66
          AK085347.1 - 2 - 3 623 66
          BC120835.1 - 3 - 4 865 40129758 66
          BC137682.1 - 3 - 4 865 8860701 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001234.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159415 BC120835.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159415 BC137682.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159415 AK019639.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159415 NM_030224.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 898901 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 234466 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 816540 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692450 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 165155 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159415 BC120835.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159415 BC137682.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159415 AK019639.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159415 NM_030224.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07419603 - -
          Whole Mouse Genome G4122A A_52_P191431 BC120835.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P191431 BC137682.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P191431 AK019639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P191431 AK085347.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P191431 NM_030224.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27968930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09079996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09080003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09071271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09279427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09709984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09329708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09079994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09071274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09416109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09022456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09022451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09022462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09416104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27968932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09343979 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10379590 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379591 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379589 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379592 - -
          Affymetrix HT MG-430 PM Array Plate 1430915_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10379588 - -
          Mouse Genome 430 2.0 Array 1430915_at - -
          Mouse Genome 430 2.0 Array 1430915_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78919_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78919_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100011568.mm.1 NM_030224.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011568.mm.1 NM_030224.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011568.mm.1 TC1100001234.mm.1 MTA TC
          Mouse Expression Set 430 1430915_PM_at - -
          Mouse Expression Set 430 1430915_at - -
          Affymetrix HT Mouse Genome 430B Array 1430915_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430915_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 722483 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101580114 - -
          Illumina mouse-6 v1.0 expression beadchip 101580114 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247113 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223273 - -
          Illumina Sentrix mouse-6.v1 bead array 101580114 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101580114 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2859932 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247113 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2859932 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline