Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01309913_m1
          See other TMEM199 GE Assays ›
          Gene Symbol:
          Tmem199
          Gene Name:
          transmembrane protein 199
          Gene Aliases:
          AI316787, AI848469, R74819
          Chromosome Location:
          Chr.11: 78507055 - 78512168 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01309913_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem199
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199199.3 NP_954669.2 3 - 4 438 69
          GenBank mRNA AK156856.1 - 3 - 4 373 69
          AK158039.1 - 3 - 4 402 69
          BC050977.1 - 3 - 4 401 69
          BC147070.1 - 3 - 4 426 9056057 69
          BC147071.1 - 3 - 4 426 9056065 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003298.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem199

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27943789 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27943788 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58471 BC050977.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58471 BC064015.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58471 NM_199199.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58471 AK160676.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58471 BC050977.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58471 BC064015.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58471 NM_199199.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58471 AK160676.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 929846 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 357773 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 689819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 697546 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147562 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145852 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58471 BC050977.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58471 BC064015.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58471 NM_199199.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58471 AK160676.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58471 BC050977.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58471 BC064015.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58471 NM_199199.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58471 AK160676.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943771 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147562 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145852 - -
          Whole Mouse Genome G4122A A_52_P58471 BC050977.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58471 BC064015.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58471 NM_199199.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58471 AK160676.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58471 BC050977.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58471 BC064015.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58471 NM_199199.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58471 AK160676.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118691 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145852 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147562 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09724266 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09724272 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09724267 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09724269 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09724271 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27943788 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27943789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08993773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09524236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09724272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09400914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09035658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943757 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10388868 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388865 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388864 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388862 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388867 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388866 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388863 - -
          Affymetrix HT MG-430 PM Array Plate 1433629_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10388861 - -
          Mouse Genome 430 2.0 Array 1433629_s_at - -
          Mouse Genome 430 2.0 Array 1433629_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 195040_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 195040_at - -
          Affymetrix Mouse Clariom S Array PSR1100030420.mm.1 NM_199199.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030420.mm.1 NM_199199.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030420.mm.1 TC1100003298.mm.1 MTA TC
          Mouse Expression Set 430 1433629_PM_s_at - -
          Mouse Expression Set 430 1433629_s_at - -
          Murine Genome U74v2 Set 93799_PM_at - -
          Murine Genome U74v2 Set 93799_at - -
          Mouse Genome 430A 2.0 Array 1433629_s_at - -
          Mouse Genome 430A 2.0 Array 1433629_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1433629_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1433629_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 784260 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5080020 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005080020 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2645719 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2645719 - -
          mouse-6 v1.1 (Illumina) scl39882.5_436-S - -
          Illumina Sentrix mouse-6.v1 bead array 5080020 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39882.5_436-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5080020 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2645719 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2645719 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl39882.5_436-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0