Search
Search
| Gene Symbol: Brs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009766.3 | NP_033896.2 | 1 - 2 | 605 | 64 | |
| GenBank mRNA | AK038616.1 | - | 1 - 2 | 606 | 64 | |
| BC106961.1 | - | 1 - 2 | 586 | 40039080 | 64 | |
| BC106962.1 | - | 1 - 2 | 586 | 40039081 | 64 | |
Set Membership: |
|