Search
Search

4930564B18Rik| Gene Symbol: 4930564B18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029230.1 | NP_083506.1 | 3 - 4 | 465 | 61 | |
| GenBank mRNA | AK016216.1 | - | 3 - 4 | 466 | 61 | |
Set Membership: |
|