Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01310735_m1
          See other LCP1 GE Assays ›
          Gene Symbol:
          Lcp1
          Gene Name:
          lymphocyte cytosolic protein 1
          Gene Aliases:
          AW536232, D14Ertd310e, LCP-1, Pls2, pp65
          Chromosome Location:
          Chr.14: 75131105 - 75230842 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01310735_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lcp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001247984.1 NP_001234913.1 2 - 3 677 86
          NM_008879.4 NP_032905.2 3 - 4 425 86
          GenBank mRNA AK030806.1 - 2 - 3 131 86
          AK030987.1 - 2 - 3 298 86
          AK030996.1 - 2 - 3 159 86
          AK089922.1 - 2 - 3 147 86
          AK149790.1 - 2 - 3 247 86
          AK151155.1 - 2 - 3 131 86
          AK151542.1 - 2 - 3 145 86
          AK151720.1 - 2 - 3 145 86
          AK152668.1 - 2 - 3 145 86
          AK152891.1 - 2 - 3 144 86
          AK152902.1 - 2 - 3 144 86
          AK153082.1 - 2 - 3 152 86
          AK167261.1 - 2 - 3 154 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001054.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lcp1

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          T cell activation involved in immune response
          response to wounding
          protein kinase A signaling
          cell migration
          extracellular matrix disassembly
          regulation of intracellular protein transport
          actin filament bundle assembly
          positive regulation of podosome assembly
          actin binding
          calcium ion binding
          protein binding
          identical protein binding
          metal ion binding
          actin filament binding
          GTPase binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1400008414.mm.1 NM_001247984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008414.mm.1 NM_001247984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008414.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008417.mm.1 NM_008879.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008417.mm.1 NM_008879.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008417.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008422.mm.1 NM_008879.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008422.mm.1 NM_008879.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008422.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008434.mm.1 NM_001247984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008434.mm.1 NM_001247984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008434.mm.1 NM_008879.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008434.mm.1 NM_008879.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008434.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008441.mm.1 NM_001247984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008441.mm.1 NM_001247984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008441.mm.1 NM_008879.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008441.mm.1 NM_008879.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008441.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008409.mm.1 NM_001247984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008409.mm.1 NM_001247984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008409.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008413.mm.1 NM_001247984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008413.mm.1 NM_001247984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008413.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008423.mm.1 NM_008879.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008423.mm.1 NM_008879.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008423.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008429.mm.1 NM_001247984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008429.mm.1 NM_001247984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008429.mm.1 NM_008879.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008429.mm.1 NM_008879.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008429.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008431.mm.1 NM_001247984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008431.mm.1 NM_001247984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008431.mm.1 NM_008879.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008431.mm.1 NM_008879.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008431.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008438.mm.1 NM_001247984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008438.mm.1 NM_001247984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008438.mm.1 NM_008879.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008438.mm.1 NM_008879.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008438.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008410.mm.1 NM_001247984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008410.mm.1 NM_001247984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008410.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008412.mm.1 NM_001247984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008412.mm.1 NM_001247984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008412.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008445.mm.1 NM_001247984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008445.mm.1 NM_001247984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008445.mm.1 NM_008879.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008445.mm.1 NM_008879.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008445.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008436.mm.1 NM_001247984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008436.mm.1 NM_001247984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008436.mm.1 NM_008879.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008436.mm.1 NM_008879.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008436.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008416.mm.1 NM_008879.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008416.mm.1 NM_008879.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008416.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008442.mm.1 NM_001247984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008442.mm.1 NM_001247984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008442.mm.1 NM_008879.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008442.mm.1 NM_008879.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008442.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008443.mm.1 NM_001247984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008443.mm.1 NM_001247984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008443.mm.1 NM_008879.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008443.mm.1 NM_008879.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008443.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008444.mm.1 NM_001247984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008444.mm.1 NM_001247984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008444.mm.1 NM_008879.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008444.mm.1 NM_008879.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008444.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008439.mm.1 NM_001247984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008439.mm.1 NM_001247984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008439.mm.1 NM_008879.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008439.mm.1 NM_008879.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008439.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008440.mm.1 NM_001247984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008440.mm.1 NM_001247984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008440.mm.1 NM_008879.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008440.mm.1 NM_008879.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008440.mm.1 TC1400001054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008432.mm.1 NM_001247984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008432.mm.1 NM_001247984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008432.mm.1 NM_008879.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008432.mm.1 NM_008879.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008432.mm.1 TC1400001054.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 927248 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline