Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01310818_m1
          See other PFN4 GE Assays ›
          Gene Symbol:
          Pfn4
          Gene Name:
          profilin family, member 4
          Gene Aliases:
          2900024P18Rik
          Chromosome Location:
          Chr.12: 4768913 - 4778808 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01310818_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pfn4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028376.3 NP_082652.1 2 - 3 1116 85
          GenBank mRNA AK013595.1 - 2 - 3 1118 85
          AY495949.1 - 3 - 4 659 85
          BC094614.1 - 2 - 3 1389 85
          BC116888.1 - 2 - 3 960 40126207 85
          BC116890.1 - 3 - 4 547 40126209 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000032.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pfn4

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          lipid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28202320 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P139629 NM_028376.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P139629 AK013595.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P139629 AY495949.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P139629 BC094614.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P139629 BC116888.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P139629 NM_028376.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P139629 AK013595.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P139629 AY495949.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P139629 BC094614.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P139629 BC116888.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731366 AK009291.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731366 AK122548.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731366 AK147464.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731366 AK147593.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731366 AK155296.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731366 AY178784.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731366 AY305656.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731366 AY305657.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731366 AY305658.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731366 BC094373.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731366 NM_001001602.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731366 NM_001114124.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731366 NM_001114125.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853037 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 366998 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 637396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746042 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 643431 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 29514 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140628 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P139629 NM_028376.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P139629 AK013595.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P139629 AY495949.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P139629 BC094614.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P139629 BC116888.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P139629 NM_028376.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P139629 AK013595.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P139629 AY495949.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P139629 BC094614.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P139629 BC116888.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P139629 NM_028376.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P139629 AK013595.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P139629 AY495949.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P139629 BC094614.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P139629 BC116888.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P139629 NM_028376.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P139629 AK013595.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P139629 AY495949.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P139629 BC094614.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P139629 BC116888.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202342 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140628 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02450582 - -
          Whole Mouse Genome G4122A A_52_P139629 NM_028376.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P139629 AK013595.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P139629 AY495949.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P139629 BC094614.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P139629 BC116888.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P139629 NM_028376.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P139629 AK013595.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P139629 AY495949.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P139629 BC094614.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P139629 BC116888.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131932 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140628 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09906337 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09906333 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09906332 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28202320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10116915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09906340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10104769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09906333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10338965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202337 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10394334 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394339 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394333 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394337 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394341 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394336 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394340 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394335 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394343 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394338 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394342 - -
          Affymetrix HT MG-430 PM Array Plate 1428665_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10394331 - -
          Mouse Genome 430 2.0 Array 1428665_PM_at - -
          Mouse Genome 430 2.0 Array 1428665_at - -
          Affymetrix Mouse Clariom S Array PSR1200000435.mm.1 NM_028376.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000435.mm.1 NM_028376.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000435.mm.1 TC1200000032.mm.1 MTA TC
          Mouse Expression Set 430 1428665_at - -
          Mouse Expression Set 430 1428665_PM_at - -
          Murine Genome U74v2 Set 166067_PM_f_at - -
          Murine Genome U74v2 Set 166067_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1428665_at - -
          Affymetrix HT Mouse Genome 430B Array 1428665_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 409394 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5860170 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2618400 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2618400 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2618398 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline