Search
Search

Mfsd2b| Gene Symbol: Mfsd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033488.2 | NP_001028660.2 | 2 - 3 | 216 | 67 | |
| GenBank mRNA | AK133061.1 | - | 2 - 3 | 217 | 67 | |
| AK156138.1 | - | 3 - 4 | 368 | 67 | ||
| AK162620.1 | - | 2 - 3 | 215 | 67 | ||
| AK171660.1 | - | 2 - 3 | 213 | 67 | ||
| AK172423.1 | - | 2 - 3 | 217 | 67 | ||
| BC145584.1 | - | 2 - 3 | 215 | 67 | ||
Set Membership: |
|