Search
Search

Ankrd13c| Gene Symbol: Ankrd13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013806.1 | NP_001013828.1 | 3 - 4 | 637 | 73 | |
| GenBank mRNA | AK135906.1 | - | 4 - 5 | 373 | 73 | |
| AK157542.1 | - | 865 | 73 | |||
| BC065088.1 | - | 3 - 4 | 650 | 73 | ||
| BC138991.1 | - | 3 - 4 | 645 | 8862013 | 73 | |
| BC138992.1 | - | 4 - 5 | 624 | 8862014 | 73 | |
| BQ562764.1 | - | 3 - 4 | 186 | 73 | ||
Set Membership: |
|