Search
Search

Fam174b| Gene Symbol: Fam174b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162532.1 | NP_001156004.1 | 1 - 2 | 522 | 68 | |
| GenBank mRNA | BC034069.1 | - | 1 - 2 | 406 | 68 | |
| BC110691.1 | - | 1 - 2 | 237 | 68 | ||
Set Membership: |
|