Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01311160_m1
          See other H60A GE Assays ›
          Gene Symbol:
          H60a
          Gene Name:
          histocompatibility 60a
          Gene Aliases:
          H60
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01311160_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H60a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010400.2 NP_034530.1 740 66
          GenBank mRNA AF084643.1 - 549 66
          AK166223.1 - 697 66
          AK169909.1 - 742 66
          AK170137.1 - 713 66

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus Inventoried

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene H60a

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of immune response to tumor cell
          natural killer cell activation
          positive regulation of interferon-gamma production
          positive regulation of natural killer cell activation
          susceptibility to natural killer cell mediated cytotoxicity
          positive regulation of macrophage activation
          positive regulation of nitric oxide biosynthetic process
          cellular response to organic cyclic compound
          natural killer cell lectin-like receptor binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076680 AK169909.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076680 NM_010400.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076680 AK169909.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076680 NM_010400.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P335146 AK166223.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P335146 AK169909.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P335146 AK170137.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P335146 NM_010400.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P335146 AF084643.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335146 AK166223.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335146 AK169909.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335146 AK170137.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335146 NM_010400.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335146 AF084643.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P187898 AK166039.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P187898 AK166154.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P187898 AK171185.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P187898 AK083313.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P187898 AK083339.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119516 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10362101 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362100 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608655 - -
          Affymetrix HT MG-430 PM Array Plate 1457769_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440145_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10608655 - -
          Affymetrix Mouse Gene 1.1 ST Array 10362097 - -
          Mouse Genome 430 2.0 Array 1457769_PM_at - -
          Mouse Genome 430 2.0 Array 1439343_at - -
          Mouse Genome 430 2.0 Array 1440188_at - -
          Mouse Genome 430 2.0 Array 1439343_PM_at - -
          Mouse Genome 430 2.0 Array 1457769_at - -
          Mouse Genome 430 2.0 Array 1440145_PM_at - -
          Mouse Genome 430 2.0 Array 1440145_at - -
          Mouse Genome 430 2.0 Array 1440188_PM_at - -
          Mouse Expression Set 430 1457769_PM_at - -
          Mouse Expression Set 430 1440145_PM_at - -
          Mouse Expression Set 430 1457769_at - -
          Mouse Expression Set 430 1440145_at - -
          Murine Genome U74v2 Set 140664_r_at - -
          Murine Genome U74v2 Set 140664_PM_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1440145_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440145_at - -
          Affymetrix HT Mouse Genome 430B Array 1457769_at - -
          Affymetrix HT Mouse Genome 430B Array 1457769_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2645333 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2645333 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline